Learn More
Thermo Scientific™ Oxoid™ Microbact™ Staph Fast Blue Reagent Set
Catalog No. MB1588A Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
2 x 10mL Bottles
Catalog No. Quantity
MB1588A 2 x 10mL Bottles
1 options
This item is not returnable. View return policy
Catalog No. MB1588A Supplier Thermo Scientific™ Supplier No. MB1588A
Add to Cart
Add to Cart
This item is not returnable. View return policy
Simplify the identification of staphylococci, including staphylococcus aureus and coagulase-negative staphylococci, with the Thermo Scientific™ Oxoid™ Microbact™ Staph Fast Blue Reagent Set.
Simplify the identification of staphylococci, including staphylococcus aureus and coagulase-negative staphylococci (CNS), with the Thermo Scientific™ Oxoid™ Microbact™ Staph Fast Blue Reagent Set. This set is used in the Beta-galactosidase test for fast, reliable and easy-to-read results.
Used in the Beta-galactosidase test, this reagent set allows for quick, reliable and easy identification of staphylococci, including staphylococcus aureus and coagulase-negative staphylococci (CNS).
Not all products are available for sale in all territories. Please inquire.
Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products with a comprehensive line of media and diagnostic products, providing a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY
Specifications
Description Microbact Staph Fast Blue Reagent Set
Detectable Analytes Staphylococcus species
Type Reagent Set for Staphylococcus aureus Test Kit
Packaging Type 1 Box
Quantity 2 x 10mL Bottles
Target Organism Class Staphylococcus aureus