Learn More
Description
Oxoid Modified Karmali Selective Supplement is for a blood-free selective medium for the isolation of Campylobacter jejuni and Campylobacter coli when incubated at 42° C.
- Alternative supplement containing amphotericin B as a replacement for cycloheximide
- Add to Campylobacter Agar Base (Karmali), Part No. CM0935B
- Each vial supplements 500mL of medium
Specifications
Specifications
| Description | Modified Karmali Selective Supplement |
| Form | Freeze-dried |
| Product Type | Selective Supplement |
| Quantity | 10/Pk. |
| Yield | Each vial for 500mL medium |
| Species | Karmali |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.