Learn More
Description
Oxoid MUG Supplement (50mg) Fluorescent agent is used for the detection of E. coli.
- Contains 4-methylumbelliferyl-β-D-glucuronide (MUG)
- Each vial supplements 500mL of medium
Specifications
Specifications
| Description | MUG Supplement |
| Form | Freeze-dried |
| Product Type | Microbiology Media |
| Quantity | 10 x 50 mg |
| Yield | Yield will be dependent upon formulation; follow the guidelines for your specific product. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.