Learn More
Description
Oxoid Mycoplasma Supplement G is for the isolation of mycoplasmas.
- Add to Mycoplasma Pneumoniae Agar, Part No. CM0401B or CM0403B
- Each vial supplements 80mL of medium
Specifications
Specifications
| Description | Mycoplasma Supplement G |
| Form | Freeze-dried |
| Product Type | Microbiology Media |
| Quantity | 10/Pk. |
| Yield | Each vial for 80mL medium |
| Species | Mycoplasma species |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.