Learn More
Thermo Scientific™ Oxoid™ Neomycin Selective Supplement
Catalog No. SR0163H Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 x 150 mg
Catalog No. Quantity
SR0163H 10 x 150 mg
1 options
This item is not returnable. View return policy
Catalog No. SR0163H Supplier Thermo Scientific™ Supplier No. SR0163H
Add to Cart
Add to Cart
This item is not returnable. View return policy
The Oxoid Neomycin Selective Supplement (150mg/vial) allows the growth of clostridia and other anaerobes.
Oxoid Neomycin Selective Supplement (150mg/vial) allows the growth of clostridia and other anaerobes.
  • Add to Blood Agar, Wilkins-Chalgren Anaerobe Agar, Part No. CM0619B, or Columbia Blood Agar, Part No. CM0331B
  • Each vial supplements 2L of medium
TRUSTED_SUSTAINABILITY
Specifications
Description Neomycin Selective Supplement
Form Freeze-dried
Product Type Selective Supplement
Quantity 10 x 150 mg
Yield Each vial for 2L medium