Learn More
Thermo Scientific™ Oxoid™ Nitrocefin Solution
Catalog No. SR0112C Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 Vial Set/Pk.
Catalog No. Quantity
SR0112C 10 Vial Set/Pk.
1 options
This item is not returnable. View return policy
Catalog No. SR0112C Supplier Thermo Scientific™ Supplier No. SR0112C
Add to Cart
Add to Cart
This item is not returnable. View return policy
The Thermo Scientific™ Oxoid Nitrocefin is used for detection of beta-lactamase production.
Thermo Scientific™ Oxoid Nitrocefin is used for detection of beta-lactamase production.
  • Reconstitutes to 500μg/mL
  • Contents: 5 vials lyophilized nitrocefin (1mg) and 5 vials rehydrating fluid (1.9mL)
TRUSTED_SUSTAINABILITY
Specifications
Description Nitrocefin
Format Vial
Includes 5 vials lyophilized nitrocefin (1mg) and 5 vials rehydrating fluid (1.9mL)
Packaging Type 2 x 5 Vials
Quantity 10 Vial Set/Pk.