Learn More
Description
Thermo Scientific™ Oxoid™ ONE Broth-Salmonella Supplement is for the selective enrichment of Salmonella spp. from food samples in 24 hours in a single enrichment step.
- Add to ONE Broth-Salmonella Part No. CM1091B
- Each vial supplements 500mL of medium
Specifications
Specifications
| Description | ONE Broth-Salmonella Supplement |
| Form | Freeze-dried |
| Product Type | Microbiology Media |
| Quantity | 10/Pk. |
| Yield | Each vial for 225mL medium |
| Species | Salmonella species |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.