Learn More
Thermo Scientific™ Oxoid™ PBP2' Latex Agglutination Test Kit
GSA_VA
Catalog No. R65900 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 Tests/Kit
Catalog No. Quantity
R65900 50 Tests/Kit
1 options
This item is not returnable. View return policy
Catalog No. R65900 Supplier Thermo Scientific™ Supplier No. DR0900A
 Restricted Item
Add to Cart
Add to Cart
This item is not returnable. View return policy
For detection of PBP2' and identification of methicillin resistance in Staphylococcus aureus (MRSA) and coagulase-negative Staphylococci.
Thermo Scientific™ Oxoid PBP2' Latex Agglutination Test is a rapid latex slide agglutination test for detection of PBP2' as an aid in identifying methicillin resistance in both Staphylococcus aureus (MRSA) and coagulase-negative Staphylococci.
Ordering Alerts:
50 tests/kit.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Latex Agglutination Test Kit
For Use With (Application) Healthcare
Quantity 50 Tests/Kit