Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Oxoid™ Potassium Tellurite

Catalog No. OXSR0030J Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 x 2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OXSR0030J 10 x 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. OXSR0030J Supplier Thermo Scientific™ Supplier No. SR0030J
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Oxoid Potassium Tellurite (3.5%) is a solution for addition to culture media.

Oxoid Potassium Tellurite (3.5%) is a solution for addition to culture media.

Ordering Information:

Supplied 2mL/vial

TRUSTED_SUSTAINABILITY

Specifications

Description Potassium Tellurite
Form Liquid
Product Type Microbiology Media
Quantity 10 x 2 mL
Yield Yield will be dependent upon formulation; follow the guidelines for your specific product.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.