Description
Maintain a low moisture environment when storing AST products for manual Antimicrobial Susceptibility Testing with Thermo Scientific™ Oxoid™ Replacement Desiccant (Dispenser).
One of the simplest and most widely used methods for susceptibility testing, disc diffusion is extremely flexible, offering a wide variety of antibiotics and concentrations which can easily be changed as required.
Disc diffusion not only categorizes resistant, intermediate and sensitive organisms through quantitative results but also provides a visual indication of the following:- Inoculum level
- Presence of contamination
- Resistant mutants
- Beta-lactamase activity
- Antagonism and synergy between antibiotics
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Specifications
| Description | Oxoid Replacement Desiccant (Dispenser) |
| Product Type | Replacement Desiccant |