Description
Oxoid RPF Supplement (Rabbit Plasma Fibrinogen) is used for the isolation and enumeration of coagulase positive staphylococci from food.
- Use only with Baird-Parker Agar Base, RPF, Part No. CM0961B
- Each vial supplements 100mL of medium
Specifications
Specifications
| Description | RPF Supplement |
| Form | Freeze-dried |
| Product Type | Microbiology Media |
| Quantity | 10/Pk. |