Description
Maintain a low moisture environment while safely and conveniently storing unopened Antimicrobial Susceptibility Testing discs in the Thermo Scientific™ Oxoid™ Cartridge Container With Desiccant.
disc diffusion not only categorizes resistant, intermediate and sensitive organisms through quantitative results but also provides a visual indication of the following:
- Inoculum level
- Presence of contamination
- Resistant mutants
- Beta-lactamase activity
- Antagonism and synergy between antibiotics
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Specifications
| Description | Oxoid Cartridge Container With Desiccant |
| Product Type | Cartridge Container with Desiccant |
| Quantity | 1/Pk. |