Description
Thermo Scientific™ Oxoid SPS Disc is used for presumptive identification of Peptostreptococcus anaerobius.
Specifications
Specifications
| Description | SPS Disk |
| Detectable Analytes | P. anaerobius |
| Format | Susceptibility Disk |
| Type | SPS Disk |
| Quantity | 50 Disks/Cartridge |