Learn More
Thermo Scientific™ Oxoid™ STA Selective Supplement
Catalog No. SR0162E Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10/Pk.
Catalog No. Quantity
SR0162E 10/Pk.
1 options
This item is not returnable. View return policy
Catalog No. SR0162E Supplier Thermo Scientific™ Supplier No. SR0162E
Add to Cart
Add to Cart
This item is not returnable. View return policy
The Oxoid STA Selective Supplement (Supplement without actidione) is used for the isolation of Brochothrix thermosphacta from food samples.
Oxoid STA Selective Supplement (Supplement without actidione) is used for the isolation of Brochothrix thermosphacta from food samples.
  • Add to STAA Agar Base, Part No. CM0881B
  • Each vial supplements 500mL of medium
TRUSTED_SUSTAINABILITY
Specifications
Description STA Selective Supplement
Form Freeze-dried
Product Type Selective Supplement
Quantity 10/Pk.
Yield Each vial for 500mL medium