Description
Oxoid STAA Selective Supplement is used for the isolation of Brochothrix thermosphacta from food samples.
- Add to STAA Agar Base, Part No. CM0881B
- Each vial supplements 500mL of medium
Specifications
Specifications
| Description | STAA Selective Supplement |
| Form | Freeze-dried |
| Product Type | Selective Supplement |
| Quantity | 10/Pk. |
| Yield | Each vial for 500mL medium |