Learn More
Thermo Scientific™ Oxoid™ Trimethoprim/Sulfamethoxazole 1:19 Antimicrobial Susceptibility discs, 25μg
GSA_VA
Catalog No. CT0052B Shop All Thermo Scientific Products
Change view
Click to view available options
Concentration:
25 μg
Catalog No. Concentration
CT0052B 25 μg
1 options
This item is not returnable. View return policy
Catalog No. CT0052B Supplier Thermo Scientific™ Supplier No. CT0052B
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Determine the susceptibility of microorganisms using manual AST methods and a full range of high-quality, easy-to-use antimicrobial susceptibility testing discs.
Manually determine the antibiotic susceptibility of microorganisms using Antimicrobial Susceptibility Testing (AST) methods in conjunction with reliable, easy-to-use Thermo Scientific™ Oxoid™ Trimethoprim/Sulfamethoxazole 1:19 Antimicrobial Susceptibility discs.
Ease-of-use is provided by shatterproof spring-loaded cartridges, easy-to-read labeling and end-of-cartridge indicators.
disc diffusion not only categorizes resistant, intermediate and sensitive organisms through quantitative results but also provides a visual indication of the following:
  • Inoculum level
  • Presence of contamination
  • Resistant mutants
  • Beta-lactamase activity
  • Antagonism and synergy between antibiotics
Each cartridge is individually sealed with a desiccant to maintain a moisture level of <2%, ensuring long term stability.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Antimicrobial Agents Trimethoprim/Sulfamethoxazole
Certifications/Compliance Complies with CLSI, DIN, SFM guidelines
Concentration 25 μg
Description Oxoid Trimethoprim/Sulfamethoxazole 1:19 Antimicrobial Susceptibility Discs
Antibiotic Code SXT
Product Type Antimicrobial Susceptibility Discs
Test Type Antimicrobial Susceptibility
No. per Pack 5 Cartridges/Pk.
Packaging Type 50 Discs/Cartridge
Quantity 50 Discs/Cartridge