Learn More
Description
Thermo Scientific™ Oxoid Vancomycin Supplement is for addition to selective media for the isolation of Vancomycin Resistant Enterococci (VRE) and High Level Aminoglycoside Resistant Enterococci (HLARE) from clinical specimens.
- Each vial supplements 500mL of medium
Specifications
Specifications
| Description | Oxoid™ Vancomycin Supplement |
| Form | Powder |
| Product Type | Microbiology Media |
| Packaging Type | 5 mg/Vial |
| Quantity | 10/Pk. |
| Species | Vancomycin-resistant Enterococci (VRE), High-level Aminoglucoside-resistant Enterococci (HLARE) |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.