Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Oxoid™ VCAT Selective Supplement

Catalog No. SR0104B Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 mL of Base
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SR0104B 500 mL of Base
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. SR0104B Supplier Thermo Scientific™ Supplier No. SR0104B
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used for the preparation of New York City Medium for the isolation of Neisseria gonorrhoeae.

Thermo Scientific™ Oxoid VCAT Selective Supplement is used for the preparation of New York City Medium for the isolation of Neisseria gonorrhoeae.

Add to GC Agar Base, Part No. CM0367B
TRUSTED_SUSTAINABILITY

Specifications

Description VCAT, 10 Vials, each supplements 500mL of base
Form Liquid
Product Type Selective Supplement
Quantity 500 mL of Base
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.