Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Oxoid™ VCNT Selective Supplement

Catalog No. SR0091E
Change view
Click to view available options
Yield:
Each vial for 2L medium
Each vial for 500mL medium
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Yield
SR0091E Each vial for 500mL medium
SR0091H Each vial for 2L medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. SR0091E Supplier Thermo Scientific™ Supplier No. SR0091E
Add to Cart
Add to Cart
This item is not returnable. View return policy

The Oxoid VCNT Selective Supplement is used for the preparation of Thayer Martin Medium for the isolation of pathogenic Neisseria spp.

Oxoid VCNT Selective Supplement is used for the preparation of Thayer Martin Medium for the isolation of pathogenic Neisseria spp.

  • Add to GC Agar Base, Part No. CM0367B
TRUSTED_SUSTAINABILITY

Specifications

Description VCNT Selective Supplement
Form Freeze-dried
Product Type Selective Supplement
Quantity 10/Pk.
Yield Each vial for 500mL medium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.