Learn More
Description
Oxoid Vitox Supplement is used for the preparation of Thayer Martin Medium and New York City Medium for the isolation of Neisseria spp.
- Contents: Vitox Supplement (5 vials) plus reconstitution fluid (5 vials)
- Add to GC Agar Base, Part No. CM0367B
Ordering Information:
- Each vial of Vitox plus reconstitution fluid supplements 500mL of medium
Includes:
- Five vials of Vitox plus five vials of reconstitution fluid
Specifications
Specifications
| Description | Vitox Supplement |
| Form | Freeze-dried |
| Product Type | Microbiology Media |
| Packaging Type | 2 x 5/Pk. |
| Quantity | 5 x Freeze dried vial + 5 x Rehydration fluid |
| Yield | Each vial for 2L medium |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.