Learn More
Thermo Scientific™ Oxoid™ X Factor Discs
Catalog No. DD0003B Shop All Thermo Scientific Products
Change view
Click to view available options
Packaging Type:
1 Cart./Pk.
5 Cart./Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Type
DD0003B 5 Cart./Pk.
R65016 1 Cart./Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. DD0003B Supplier Thermo Scientific™ Supplier No. DD0003B
Add to Cart
Add to Cart
This item is not returnable. View return policy
Perform identification of Haemophilus spp. with Thermo Scientific™ Oxoid X Factor Discs.
Perform identification of Haemophilus spp. with Thermo Scientific™ Oxoid X Factor Discs.
Ordering Alerts:
50 Discs/Cartridge
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage -10°C to -20°C
Description X Factor Discs
Detectable Analytes Haemophilus species
Type X Factor Disc
Packaging Type 5 Cart./Pk.
Quantity 50 Disks/Cartridge