Learn More
Medchemexpress LLC Oxolamine citrate | 1949-20-8 | MFCD00083457 | 99.9% | 500 MG
SDP
Catalog No. 5000411588
Encompass_Preferred
Oxolamine citrate is an orally active antitussive that can inhibit CYP2B1/2. It exhibits anti-inflammatory effects on the respiratory organs. This product increases the AUC of Warfarin and prolongs its terminal half-life. It is intended for use in respiratory disease research.
  • Inhibits CYP2B1/2
  • Exhibits anti-inflammatory effects on respiratory organs
  • Increases the AUC of Warfarin
  • Prolongs Warfarin's terminal half-life
  • Reduces plasma protein binding of Clarithromycin in fresh rat plasma
  • Protects pulmonary parenchyma from emphysema in animal models
Catalog No. 50-004-11588 Supplier Medchemexpress LLC Supplier No. HYB1042500MG
Oxolamine citrate is an orally active antitussive that can inhibit CYP2B1/2. It exhibits anti-inflammatory effects on the respiratory organs. This product increases the AUC of Warfarin and prolongs its terminal half-life. It is intended for use in respiratory disease research.
  • Inhibits CYP2B1/2
  • Exhibits anti-inflammatory effects on respiratory organs
  • Increases the AUC of Warfarin
  • Prolongs Warfarin's terminal half-life
  • Reduces plasma protein binding of Clarithromycin in fresh rat plasma
  • Protects pulmonary parenchyma from emphysema in animal models