Learn More
Selleck Chemical LLC Oxolamine Citrate
SDP
Catalog No. 5000003999
Encompass_Preferred
Oxolamine (SKF-9976 AF-438) citrate a cough suppressant is an inhibitor of CYP2B1/2 Oxolamine citrate also exhibits anti-inflammatory effect
Catalog No. 50-000-03999 Supplier Selleck Chemical LLC Supplier No. S643725MG
Oxolamine (SKF-9976 AF-438) citrate a cough suppressant is an inhibitor of CYP2B1/2 Oxolamine citrate also exhibits anti-inflammatory effect