Learn More
Medchemexpress LLC Oxolamine citrate (SKF-9976 citrate) | 1949-20-8 | 99.9% | 1 ML
SDP
Catalog No. 5000412442
Encompass_Preferred
Oxolamine citrate (SKF-9976 citrate) is an orally active antitussive that can inhibit CYP2B1/2 and has anti-inflammatory effects on the respiratory organs of guinea pigs. It increases the AUC of Warfarin and prolongs its terminal half-life. It can be used in respiratory disease research.
  • Orally active antitussive
  • Inhibits CYP2B1/2
  • Has anti-inflammatory effects on the respiratory organs of guinea pigs
  • Increases the AUC of Warfarin and prolongs its terminal half-life
  • Can be used in respiratory disease research
Catalog No. 50-004-12442 Supplier Medchemexpress LLC Supplier No. HYB104210MM/1ML
Oxolamine citrate (SKF-9976 citrate) is an orally active antitussive that can inhibit CYP2B1/2 and has anti-inflammatory effects on the respiratory organs of guinea pigs. It increases the AUC of Warfarin and prolongs its terminal half-life. It can be used in respiratory disease research.
  • Orally active antitussive
  • Inhibits CYP2B1/2
  • Has anti-inflammatory effects on the respiratory organs of guinea pigs
  • Increases the AUC of Warfarin and prolongs its terminal half-life
  • Can be used in respiratory disease research