Learn More
Research Products International Corp Oxolinic Acid 5 Grams
SDP
Catalog No. 502388425
Encompass_Preferred
Oxolinic acid is an antibiotic in the quinolone class.
It works by inhibiting the bacterial DNA gyrase enzyme, which is essential for DNA replication and repair in bacteria. This disruption of DNA processes ultimately leads to the death of the bacteria.
Catalog No. 50-238-8425 Supplier Research Products International Corp Supplier No. O214505.0
Oxolinic acid is an antibiotic in the quinolone class.
It works by inhibiting the bacterial DNA gyrase enzyme, which is essential for DNA replication and repair in bacteria. This disruption of DNA processes ultimately leads to the death of the bacteria.