Learn More
Medchemexpress LLC Oxolinic acid-d5 | 1189890-98-9 | 98.0% | 1 MG
SDP
Catalog No. 5000407505
Encompass_Preferred
Oxolinic acid-d5 is the deuterium-labeled form of Oxolinic acid, an antibiotic effective against both Gram-negative and Gram-positive bacteria. It is used in research for acute and chronic urinary tract infections. This compound functions as a DNA/RNA synthesis inhibitor and a dopamine uptake inhibitor, stimulating locomotor effects in mice. It is intended for research use only.
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Effective against Gram-negative and Gram-positive bacteria
  • Inhibits DNA/RNA synthesis
  • Acts as a dopamine uptake inhibitor
  • Stimulates locomotor effects in mice
Catalog No. 50-004-07505 Supplier Medchemexpress LLC Supplier No. HYB1002S1MG
Oxolinic acid-d5 is the deuterium-labeled form of Oxolinic acid, an antibiotic effective against both Gram-negative and Gram-positive bacteria. It is used in research for acute and chronic urinary tract infections. This compound functions as a DNA/RNA synthesis inhibitor and a dopamine uptake inhibitor, stimulating locomotor effects in mice. It is intended for research use only.
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Effective against Gram-negative and Gram-positive bacteria
  • Inhibits DNA/RNA synthesis
  • Acts as a dopamine uptake inhibitor
  • Stimulates locomotor effects in mice