Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Oxolinic acid (Standard) | 14698-29-4 | 97.6% | 25 MG
SDP

Catalog No. 5000450847
Encompass_Preferred

Oxolinic acid (Standard) is an analytical standard for research and analytical applications. It is an antibiotic effective against Gram-negative and Gram-positive bacteria, used in research for acute and chronic urinary tract infections. This compound inhibits DNA/RNA synthesis and dopamine uptake, stimulating locomotor effects in mice.

  • Analytical standard for research and analytical applications
  • Effective against Gram-negative and Gram-positive bacteria
  • Used in research for acute and chronic urinary tract infections
  • Acts as a DNA/RNA synthesis inhibitor
  • Acts as a dopamine uptake inhibitor, stimulating locomotor effects in mice
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
  • Molecular weight is 261.23
  • Formula is C13H11NO5
  • Appears as a white to off-white solid

Catalog No. 50-004-50847 Supplier Medchemexpress LLC Supplier No. 25MGHYB1002R
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Oxolinic acid (Standard) is an analytical standard for research and analytical applications. It is an antibiotic effective against Gram-negative and Gram-positive bacteria, used in research for acute and chronic urinary tract infections. This compound inhibits DNA/RNA synthesis and dopamine uptake, stimulating locomotor effects in mice.

  • Analytical standard for research and analytical applications
  • Effective against Gram-negative and Gram-positive bacteria
  • Used in research for acute and chronic urinary tract infections
  • Acts as a DNA/RNA synthesis inhibitor
  • Acts as a dopamine uptake inhibitor, stimulating locomotor effects in mice
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
  • Molecular weight is 261.23
  • Formula is C13H11NO5
  • Appears as a white to off-white solid

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.