Learn More
Medchemexpress LLC Oxolinic acid (Standard) | 14698-29-4 | 97.6% | 250 MG
SDP
Catalog No. 5000406166
Encompass_Preferred
Oxolinic acid is an antibiotic effective against both Gram-negative and Gram-positive bacteria. It can be used for research on acute and chronic urinary tract infections. It inhibits DNA/RNA synthesis and acts as a dopamine uptake inhibitor, stimulating locomotor effects in mice.
  • Antibiotic against both gram-negative and gram-positive bacteria
  • Can be used for the research of acute and chronic urinary tract infections
  • DNA/RNA synthesis inhibitor
  • Acts as a dopamine uptake inhibitor and stimulates locomotor effect in mice
Catalog No. 50-004-06166 Supplier Medchemexpress LLC Supplier No. 250MGHYB1002R
Oxolinic acid is an antibiotic effective against both Gram-negative and Gram-positive bacteria. It can be used for research on acute and chronic urinary tract infections. It inhibits DNA/RNA synthesis and acts as a dopamine uptake inhibitor, stimulating locomotor effects in mice.
  • Antibiotic against both gram-negative and gram-positive bacteria
  • Can be used for the research of acute and chronic urinary tract infections
  • DNA/RNA synthesis inhibitor
  • Acts as a dopamine uptake inhibitor and stimulates locomotor effect in mice