Learn More
Cayman Chemical (±)-Oxomemazine, Purity: ≥98%, CAS: 3689-50-7, 100 mg
SDP
Catalog No. 502919164
Encompass_Preferred
A phenothiazine antihistamine; protective against anaphylactic microshock in guinea pigs from 5-10 µg/kg
Catalog No. 50-291-9164 Supplier Cayman Chemical Supplier No. 39511100 MG
A phenothiazine antihistamine; protective against anaphylactic microshock in guinea pigs from 5-10 µg/kg