Learn More
Medchemexpress LLC Oxomemazine | 3689-50-7 | MFCD00868197 | 99.2% | C18H22N2O2S | 10MG
SDP
Catalog No. 5000261505
Encompass_Preferred
Oxomemazine is a phenothiazine-based histamine H1-receptor blocker with anticholinergic activity, supplied for research use. Common applications include studies of cough, antihistaminic effects, and muscarinic receptor modulation. Chemical identifiers: CAS 3689-50-7; formula C18H22N2O2S; molecular weight 330.44 g·mol⁻¹. Certificate of analysis reports purity ~99.18% (LCMS).
  • Phenothiazine-based H1-receptor blocker suitable for pharmacology research.
  • Anticholinergic properties useful for muscarinic receptor studies.
  • High purity suitable for analytical and assay applications.
  • Well-characterized chemical identifiers for database cross-reference.
  • Provided in small quantities appropriate for method development and screening.
Catalog No. 50-002-61505 Supplier Medchemexpress LLC Supplier No. HY13658710MG
Oxomemazine is a phenothiazine-based histamine H1-receptor blocker with anticholinergic activity, supplied for research use. Common applications include studies of cough, antihistaminic effects, and muscarinic receptor modulation. Chemical identifiers: CAS 3689-50-7; formula C18H22N2O2S; molecular weight 330.44 g·mol⁻¹. Certificate of analysis reports purity ~99.18% (LCMS).
  • Phenothiazine-based H1-receptor blocker suitable for pharmacology research.
  • Anticholinergic properties useful for muscarinic receptor studies.
  • High purity suitable for analytical and assay applications.
  • Well-characterized chemical identifiers for database cross-reference.
  • Provided in small quantities appropriate for method development and screening.