Learn More
Cayman Chemical (±)-Oxomemazine, Purity: ≥98%, CAS: 3689-50-7, 1 g
SDP
Catalog No. 502919223
Encompass_Preferred
A phenothiazine antihistamine; protective against anaphylactic microshock in guinea pigs from 5-10 µg/kg
Catalog No. 50-291-9223 Supplier Cayman Chemical Supplier No. 395111 G
A phenothiazine antihistamine; protective against anaphylactic microshock in guinea pigs from 5-10 µg/kg