Learn More
Medchemexpress LLC Oxonic acid -13C2,15N3 potassium salt hydrate | 1644063-37-5 | MFCD00010565 | ≥95.0% | 218.16 | C213C2H4K15N3O5 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYW6543041MG
Encompass_Preferred
Oxonic acid-13C2,15N3 potassium salt hydrate is an isotopically labeled internal standard supplied as a small, accurately weighed solid for analytical quantification. The 13C and 15N labels provide clear mass separation from unlabeled analyte, enabling precise tracer studies and quantitative workflows in NMR, GC-MS, and LC-MS. Certificate of analysis is provided for identity and purity verification.
  • Isotopically labeled with 13C and 15N for unambiguous mass spectrometric identification.
  • Suitable as an internal standard or tracer in NMR, GC-MS, and LC-MS.
  • Provided as a neat solid with exact weight packaging for precise quantitation.
  • Available in 1 mg size, with larger quantities available on request.
  • Certificate of analysis supplied for purity and identity confirmation.
Catalog No. 50-002-84031