Learn More
Cayman Chemical OxonIc AcIdpotasIum salt 25g
SDP
Catalog No. 502986820
Encompass_Preferred
A uricase inhibitor; induces embryotoxicity and nephrotoxicity in rats; a component of S-1, a mixture containing a prodrug of the antitumor agent 5-FU, that suppresses the GI toxicity of 5-FU without inhibiting its antitumor activity in rats
Catalog No. 50-298-6820 Supplier Cayman Chemical Supplier No. 2258625 G
A uricase inhibitor; induces embryotoxicity and nephrotoxicity in rats; a component of S-1, a mixture containing a prodrug of the antitumor agent 5-FU, that suppresses the GI toxicity of 5-FU without inhibiting its antitumor activity in rats