Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oxonol 805 Blue, TCI America™
SDP

Catalog No. O013125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O013125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O013125G Supplier TCI America Supplier No. O013125G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 27969-56-8
Molecular Formula C25H22N4O8S2
MDL Number MFCD00213904
Synonym 1,5-Pentamethin-4,4′C-bis[1-(p-sulfophenyl)-3-methyl-5-pyrazolone]

Specifications

Color Black-Yellow
Quantity 25 g
Formula Weight 570.59
Physical Form Crystalline Powder
Chemical Name or Material Oxonol 805 Blue
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.