Learn More
AdipoGen Oxonol V
SDP
Catalog No. 502560277
Encompass_Preferred
Chemical. CAS 61389-30-8. Formula C23H16N2O4. MW 384.38. Synthetic. Potential sensitive probe. Determination of plasma membrane potential of neutrophils generated by the Na pump.
Catalog No. 50-256-0277 Supplier AdipoGen Supplier No. CDXP0019M500
Chemical. CAS 61389-30-8. Formula C23H16N2O4. MW 384.38. Synthetic. Potential sensitive probe. Determination of plasma membrane potential of neutrophils generated by the Na pump.