Learn More
Abcam O x onol V, Fluorescent membrane indicator, 50MG
Catalog No. 502705244
Encompass_Preferred
MW 384.38 g/mol, Purity >95%. Anionic dye. Fluorescent membrane potential indicator. Carries a negative electric charge. Monitors positive membrane potentials inside vesicles. Provides estimates of transmembrane potentials.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5244 Supplier Abcam Supplier No. AB14538450MG
MW 384.38 g/mol, Purity >95%. Anionic dye. Fluorescent membrane potential indicator. Carries a negative electric charge. Monitors positive membrane potentials inside vesicles. Provides estimates of transmembrane potentials.
The product is subject to the following: Abcam Restricted Use Statement