Learn More
Cayman Chemical OxotremorIn MIodIde 50mg
SDP
Catalog No. 502980747
Encompass_Preferred
A muscarinic acetylcholine receptor agonist and KCNQ2/3 potassium channel blocker; induces phosphoinositide hydrolysis in mouse ileum isolated from wild-type, M2, M3 and M2/M3 knockout mice (EC50s = 0.36, 0.52, 1.62, and 1.48 μM, respectively); increases M4-induced inhibition of calcium currents in NG108-15 mouse neuroblastoma X rat glioma hybrid cells (EC50 = 0.14 μM); inhibits KCNQ2/3 potassium currents in Xenopus oocytes expressing M1 receptors and KCNQ2/3 potassium channels (IC50 = 1.1 μM); decreases KCNQ2/3 potassium currents in Xenopus oocytes lacking muscarinic receptors in a dose-dependent manner
Catalog No. 50-298-0747 Supplier Cayman Chemical Supplier No. 2084750 MG
A muscarinic acetylcholine receptor agonist and KCNQ2/3 potassium channel blocker; induces phosphoinositide hydrolysis in mouse ileum isolated from wild-type, M2, M3 and M2/M3 knockout mice (EC50s = 0.36, 0.52, 1.62, and 1.48 μM, respectively); increases M4-induced inhibition of calcium currents in NG108-15 mouse neuroblastoma X rat glioma hybrid cells (EC50 = 0.14 μM); inhibits KCNQ2/3 potassium currents in Xenopus oocytes expressing M1 receptors and KCNQ2/3 potassium channels (IC50 = 1.1 μM); decreases KCNQ2/3 potassium currents in Xenopus oocytes lacking muscarinic receptors in a dose-dependent manner