Learn More
Eurofins Discoverx Oxotremorine-M, 10 mg. (92-1106)
Supplier:  Eurofins Discoverx 921106
Encompass
Oxotremorine-M is synthetic parasympathomimetic drug that is a non-selective agonist of the muscarinic acetylcholine receptors (CHRM1-4). Produces symptoms of ataxia, spasms, and tremor, and possesses antipsychotic effects. Is used to induce Parkinson's-like symptoms for studies of anti-parkinsonian drugs. Provided as 10 mg of dry powder alongside reconstitution buffer. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9641