Learn More
Medchemexpress LLC HY-101372A 100mg Medchemexpress, Oxotremorine M (iodide) CAS:3854-04-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY101372A100MG
Encompass_Preferred
Medchemexpress, HY-101372A 100mg Oxotremorine M (iodide) CAS:3854-04-4 Oxotremorine M iodide is a potent and selective muscarinic acetylcholine receptor (mAChR) agonist. Oxotremorine M iodide potentiates NMDA receptors by muscarinic receptor dependent and independent mechanisms [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2103