Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Oxotrichloro[(Dimethylsulfide)Triphenylphosphine Oxide]Rhenium(V)

Catalog No. 6650961G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
66-509-61G 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 66-509-61G Supplier Sigma Aldrich Supplier No. 6650961G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Catalyst for:Enantioselective reduction of ketones and iminesMeyer-Schuster rearrangement of propargylic alcohols to +a, +a-unsaturated carbonyl compoundsSynthesis of 2-deoxy-+a glycosidesSulfurization of phosphorous(III) compounds with thiiranesReactant for synthesis of mono and dinuclear compoundsReagent for rhenium-cyclized somatostatine derivative series synthesis

Specifications

CAS 947601-98-1
Molecular Formula C46H80N2O24S2
Linear Formula C46H80N2O24S2
MDL Number MFCD08274613
Quantity 100 mg
Synonym Trichlorooxo[thiobis[methane]](triphenylphosphine oxide-kappaO)rhenium
Molecular Weight (g/mol) 1109.26 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.