Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Oxprenoate potassium | 76676-34-1 | 98.0% | 440.66 | C25H37KO4 | 1 MG
SDP

Catalog No. 5000242022
Encompass_Preferred

Oxprenoate potassium is a synthetic steroidal mineralocorticoid receptor (MR) antagonist used as a research reagent. It has been reported to accentuate dexamethasone-induced apoptosis and is supplied as a high-purity material for biochemical and cell-based assays.

  • Purity 98.0%.
  • Molecular weight 440.66.
  • Molecular formula C25H37KO4.
  • CAS number 76676-34-1.
  • Supplied as a 1 mg vial.
  • Activity: mineralocorticoid receptor antagonist.
  • Intended for in vitro research applications.

Catalog No. 50-002-42022 Supplier Medchemexpress LLC Supplier No. HY1076311MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Oxprenoate potassium is a synthetic steroidal mineralocorticoid receptor (MR) antagonist used as a research reagent. It has been reported to accentuate dexamethasone-induced apoptosis and is supplied as a high-purity material for biochemical and cell-based assays.

  • Purity 98.0%.
  • Molecular weight 440.66.
  • Molecular formula C25H37KO4.
  • CAS number 76676-34-1.
  • Supplied as a 1 mg vial.
  • Activity: mineralocorticoid receptor antagonist.
  • Intended for in vitro research applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.