Learn More
Medchemexpress LLC Oxprenolol-d7 hydrochloride | 1189649-47-5 | 98.3% | 1 MG
SDP
Catalog No. 5000411378
Encompass_Preferred
Oxprenolol-d7 (hydrochloride) is the deuterium labeled Oxprenolol hydrochloride. Oxprenolol hydrochloride is an orally bioavailable β-adrenergic receptor (β-AR) antagonist with a Ki of 7.10 nM in a radioligand binding assay using rat heart muscle.
  • Can be used as a tracer.
  • Serves as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Incorporates stable heavy isotopes as tracers for quantitation during drug development.
  • Deuteration may affect pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-11378 Supplier Medchemexpress LLC Supplier No. HYB1486S1MG
Oxprenolol-d7 (hydrochloride) is the deuterium labeled Oxprenolol hydrochloride. Oxprenolol hydrochloride is an orally bioavailable β-adrenergic receptor (β-AR) antagonist with a Ki of 7.10 nM in a radioligand binding assay using rat heart muscle.
  • Can be used as a tracer.
  • Serves as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Incorporates stable heavy isotopes as tracers for quantitation during drug development.
  • Deuteration may affect pharmacokinetic and metabolic profiles of drugs.