Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Oxprenolol-d7 hydrochloride | 1189649-47-5 | 98.3% | 308.85 | C15H17D7ClNO3 | 10 MG
SDP

Catalog No. 5000298039
Encompass_Preferred

Oxprenolol-d7 hydrochloride is a deuterium-labeled hydrochloride salt of oxprenolol used as an analytical standard and internal standard for LC-MS and other quantitative assays. Supplied at high purity in mg-scale packages, follow recommended cold storage to preserve stability.

  • Deuterium-labeled hydrochloride salt for use as an analytical/internal standard.
  • High purity (98.25%) for accurate quantitation.
  • Available in mg-scale packages to support method development.
  • Recommended storage: powder -20°C (3 years) or 4°C (2 years).
  • Molecular formula C15H17D7ClNO3 and molecular weight 308.85 for identification.

Catalog No. 50-002-98039 Supplier Medchemexpress LLC Supplier No. HYB1486S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Oxprenolol-d7 hydrochloride is a deuterium-labeled hydrochloride salt of oxprenolol used as an analytical standard and internal standard for LC-MS and other quantitative assays. Supplied at high purity in mg-scale packages, follow recommended cold storage to preserve stability.

  • Deuterium-labeled hydrochloride salt for use as an analytical/internal standard.
  • High purity (98.25%) for accurate quantitation.
  • Available in mg-scale packages to support method development.
  • Recommended storage: powder -20°C (3 years) or 4°C (2 years).
  • Molecular formula C15H17D7ClNO3 and molecular weight 308.85 for identification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.