Learn More
Medchemexpress LLC Oxprenolol (hydrochloride) | 6452-73-9 | 99.9% | 1 ML
SDP
Catalog No. 5000411683
Encompass_Preferred
Oxprenolol hydrochloride is an orally bioavailable β-adrenergic receptor (β-AR) antagonist with a Ki of 7.10 nM in a radioligand binding assay using rat heart muscle. It is for research use only.
  • Orally bioavailable β-adrenergic receptor (β-AR) antagonist
  • Ki of 7.10 nM in a radioligand binding assay using rat heart muscle
  • Lipophilic
  • Permeability rate constant of 1.54 ± 1.54x10⁻³ cm/h across abdominal human skin
  • Produces effective beta-blockade in male Wistar rats at 200 mg/kg/day orally for 3 weeks
Catalog No. 50-004-11683 Supplier Medchemexpress LLC Supplier No. HYB148610MM/1ML
Oxprenolol hydrochloride is an orally bioavailable β-adrenergic receptor (β-AR) antagonist with a Ki of 7.10 nM in a radioligand binding assay using rat heart muscle. It is for research use only.
  • Orally bioavailable β-adrenergic receptor (β-AR) antagonist
  • Ki of 7.10 nM in a radioligand binding assay using rat heart muscle
  • Lipophilic
  • Permeability rate constant of 1.54 ± 1.54x10⁻³ cm/h across abdominal human skin
  • Produces effective beta-blockade in male Wistar rats at 200 mg/kg/day orally for 3 weeks