Learn More
Aviva Systems Biology Corp OXR1 Antibody - N-terminal region (ARP57776_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57776_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against OXR1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human OXR1. Alternate names: FLJ10125, FLJ38829, FLJ41673, FLJ42450, FLJ45656, Nbla00307ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-62156