Learn More
Aviva Systems Biology Corp Oxsr1 Antibody - middle region (ARP48661_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48661_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Oxsr1. It was validated on Western Blot. . Alternate names: 2210022N24Rik, 2810422B09Rik, AI462649, AW209236, Osr1, mKIAA1101ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-285-373