Learn More
ABclonal Technology OXSR1 Rabbit pAb
SDP
Catalog No. 5000105498
Encompass_Preferred
The product of this gene belongs to the Ser/Thr protein kinase family of proteins. It regulates downstream kinases in response to environmental stress, and may play a role in regulating the actin cytoskeleton.
Catalog No. 50-001-05498 Supplier ABclonal Technology Supplier No. A15126500UL
The product of this gene belongs to the Ser/Thr protein kinase family of proteins. It regulates downstream kinases in response to environmental stress, and may play a role in regulating the actin cytoskeleton.