Learn More
Aviva Systems Biology Corp OXT Antibody - N-terminal region (ARP41829_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41829_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against OXT. It was validated on Western Blot. . Alternate names: MGC126890, MGC126892, OT, OT-NPIApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-284-958