Learn More
Eurofins Discoverx Ready-to-Assay OT Oxytocin Receptor Frozen Cells, 2 Vials of Cells. (HTS090RTA)
Catalog No. 502005847
Encompass
The Ready-to-Assay OT Oxytocin Receptor Frozen Cells are a simple, thaw-and-assay cell format for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This assay assesses ligand (e.g. oxytocin) based activation of OXTR GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses OXTR alongside endogenous high levels of a promiscuous G-protein, coupling the receptor to the calcium signaling pathway. Upon activation of OXTR, the resulting calcium activity can be fluorescently assayed. Ready-to-assay kits consist of two vials of the stably modified clonal cell line in a convenient thaw-and-use format. For detailed product information, User manual and datasheet, visit www.discoverx.com/products-applications/frozen-cells.
Catalog No. 50-200-5847 Supplier Eurofins Discoverx Supplier No. HTS090RTA
Add to Cart
Add to Cart
The Ready-to-Assay OT Oxytocin Receptor Frozen Cells are a simple, thaw-and-assay cell format for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This assay assesses ligand (e.g. oxytocin) based activation of OXTR GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses OXTR alongside endogenous high levels of a promiscuous G-protein, coupling the receptor to the calcium signaling pathway. Upon activation of OXTR, the resulting calcium activity can be fluorescently assayed. Ready-to-assay kits consist of two vials of the stably modified clonal cell line in a convenient thaw-and-use format. For detailed product information, User manual and datasheet, visit www.discoverx.com/products-applications/frozen-cells.