Learn More
Cayman Chemical OxtrIphylIn 50mg
SDP
Catalog No. 502933038
Encompass_Preferred
A choline salt form of theophylline; decreases histamine-induced contractions of isolated guinea pig trachea; increases cAMP levels in guinea pig tracheal smooth muscle cells in vitro
Catalog No. 50-293-3038 Supplier Cayman Chemical Supplier No. 2400650 MG
A choline salt form of theophylline; decreases histamine-induced contractions of isolated guinea pig trachea; increases cAMP levels in guinea pig tracheal smooth muscle cells in vitro